Anti-POT1 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB500-176C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Protection of telomeres 1
- Antigen symbol:POT1
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:25913
- Antigen synonyms:POT 1|POT-1|Protection of telomeres 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: POT1 (protection of telomeres 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human