Order Entry
Denmark
ContactUsLinkComponent
Anti-ABCA7 Rat Monoclonal Antibody (DyLight® 488) [clone: 7A1-144]
Anti-ABCA7 Rat Monoclonal Antibody (DyLight® 488) [clone: 7A1-144]
  NOVUNB400-163G
Anti-ABCA7 Rat Monoclonal Antibody (DyLight® 488) [clone: 7A1-144]
  NOVUNB400-163G

 

  • Antibody type:
    Primary
  • Antigen name:
    ATP-binding cassette, Subfamily A (ABC1), Member 7
  • Antigen symbol:
    ABCA7
  • Clonality:
    Monoclonal
  • Clone:
    7A1-144
  • Conjugation:
    DyLight® 488
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    10347
  • Antigen synonyms:
    ATP-binding Cassette 1|CERP|ABC1|ABC Transporter 1|ABC 1|ATP-binding Cassette Transporter 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: monoclonal
Clone: 7A1-144
Conjugation: DyLight® 488
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse