Anti-Niemann-Pick C1 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB400-148C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Niemann-Pick disease type C1
- Antigen symbol:NPC1
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:4864
- Antigen synonyms:NPC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Niemann-Pick C1
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: