Order Entry
Denmark
ContactUsLinkComponent
Anti-THOP1 Mouse Monoclonal Antibody (Biotin) [clone: 4D6]
Anti-THOP1 Mouse Monoclonal Antibody (Biotin) [clone: 4D6]
Katalog # NOVUNB400-146B
Supplier:  Novus Biologicals
undefined
Anti-THOP1 Mouse Monoclonal Antibody (Biotin) [clone: 4D6]
Katalog # NOVUNB400-146B
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thimet Oligopeptidase 1
  • Antigen symbol:
    THOP1
  • Clonality:
    Monoclonal
  • Clone:
    4D6
  • Conjugation:
    Biotin
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    7064
  • Antigen synonyms:
    MP78|TOP|MEPD_HUMAN|EP24.15
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: THOP1 (thimet oligopeptidase 1)
Clonality: monoclonal
Clone: 4D6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: