Anti-CD36 Rabbit Polyclonal Antibody (Biotin)
NOVUNB400-144B
- Antibody type:Primary
- Antigen name:CD36 molecule
- Antigen symbol:CD36
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:948
- Antigen synonyms:GP4|GP3B|FAT|GPIV|SCARB3|CHDS7|BDPLT10|PASIV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: