Anti-MLXIPL Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: MLXIPL (MLX interacting protein-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB400-135SS
- NOVUNB400-135
- Antigen symbol:
- ChREBP
- ChREBP
- Antigen name:
- Carbohydrate-responsive element-binding protein
- Carbohydrate-responsive element-binding protein
- Antigen synonyms:
- WBSCR14|WS-bHLH|MIO|MONDOB|Williams Beuren Syndrome Chromosome Region 14
- WBSCR14|WS-bHLH|MIO|MONDOB|Williams Beuren Syndrome Chromosome Region 14
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 51085
- 51085
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together