Anti-ABCG8 Rabbit Polyclonal Antibody (FITC (Fluorescein))
NOVUNB400-117F
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family G (WHITE), member 8
- Antigen symbol:ABCG8
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:64241
- Antigen synonyms:STSL|GBD4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 μL
- Pk:0,2 mL
Type: Primary
Antigen: ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse