Anti-SCARB1 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNB400-101C
- Antibody type:Primary
- Antigen name:Scavenger receptor class B, member 1
- Antigen symbol:SCARB1
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:949
- Antigen synonyms:SR-BI|mSR-BI|SR-B1|CD36|Cla-1|Hdlq1|Hlb398|SRBI|Srb1|D5Ertd460e|Cd36l1|Cla1|AI120173
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: