Order Entry
Denmark
ContactUsLinkComponent
 
Anti-HSPA5 Rabbit Polyclonal Antibody (DyLight® 405)
  NOVUNB300-520V
Anti-HSPA5 Rabbit Polyclonal Antibody (DyLight® 405)
  NOVUNB300-520V

 

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock 70kDa protein 5
  • Antigen symbol:
    HSPA5
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    3309
  • Antigen synonyms:
    GRP78|BIP|MIF2|HEL-S-89n
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat