Order Entry
Denmark
ContactUsLinkComponent
Anti-FXR2 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1G2]
Anti-FXR2 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1G2]
Catalog # NOVUNB300-259R
Supplier:  Novus Biologicals
Anti-FXR2 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1G2]
Catalog # NOVUNB300-259R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FMR1 autosomal homolog 2
  • Antigen symbol:
    FXR2
  • Clonality:
    Monoclonal
  • Clone:
    1G2
  • Conjugation:
    DyLight® 550
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    9513
  • Antigen synonyms:
    FMR1L2|FXR2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: FXR2 (fragile X mental retardation, autosomal homolog 2)
Clonality: monoclonal
Clone: 1G2
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse