Anti-CTDSPL Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB300-231F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Synaptonemal Complex Protein 3
- Antigen symbol:SYCP3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:50511
- Antigen synonyms:SPGF4|COR1|SCP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CTDSPL (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse