Anti-PSENEN Rabbit Polyclonal Antibody
Type: Primary
Antigen: PSENEN (presenilin enhancer gamma secretase subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
- Catalog No:
- NOVUNB300-227SS
- NOVUNB300-227
- Antigen symbol:
- PEN2
- PEN2
- Antigen name:
- Presenilin Enhancer 2 homolog
- Presenilin Enhancer 2 homolog
- Antigen synonyms:
- PEN2|MDS033|PEN-2|MSTP064
- PEN2|MDS033|PEN-2|MSTP064
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 55851
- 55851
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together