Order Entry
Denmark
ContactUsLinkComponent
 
Anti-SSTR2 Rabbit Polyclonal Antibody
Catalog # NOVUNB300-157SS
Supplier:  Novus Biologicals
Anti-SSTR2 Rabbit Polyclonal Antibody
Catalog # NOVUNB300-157SS
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Somatostatin Receptor 2
  • Antigen symbol:
    SSTR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    6752
  • Antigen synonyms:
    SSTR2|SSR2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    25 µL
  • Pk:
    25 µl

Specifications

About this item

Type: Primary
Antigen: SSTR2 (somatostatin receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat