Anti-HIST4H4 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB21-2089V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:histone cluster 4, H4
- Antigen symbol:HIST4H4
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Gene ID:121504
- Antigen synonyms:H4/p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: