Order Entry
Denmark
ContactUsLinkComponent
Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 405)
Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 405)
  NOVUNB21-1188V
Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 405)
  NOVUNB21-1188V

 

  • Antibody type:
    Primary
  • Antigen name:
    histone cluster 2, H3c
  • Antigen symbol:
    HIST2H3C
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    126961
  • Antigen synonyms:
    H3F2|H3.2|H3FM|H3|H3/M|H3FN
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 μg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: