Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB21-1188R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Histone Cluster 2, H3c
- Antigen symbol:HIST2H3C
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- ChIP:Yes
- Gene ID:126961
- Antigen synonyms:H3.3B|family 3A|H3R2me1|MGC87782|H3.3AH3F3H3F3B|histone H3.3|MGC87783|H3 histone
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: