Anti-SLC9A3 Mouse Monoclonal Antibody (Biotin) [clone: 14D5]
NOVUNB110-81529B
- Antibody type:Primary
- Antigen name:Sodium/hydrogen exchanger 3
- Antigen symbol:NHE3
- Clonality:Monoclonal
- Clone:14D5
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Gene ID:6550
- Antigen synonyms:sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: SLC9A3 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3)
Clonality: monoclonal
Clone: 14D5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: