Anti-SLC9A3 Mouse Monoclonal Antibody (Biotin) [clone: 10A8]
Catalog # NOVUNB110-74678B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sodium/hydrogen exchanger 3
- Antigen symbol:NHE3
- Clonality:Monoclonal
- Clone:10A8
- Conjugation:Biotin
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:6550
- Antigen synonyms:sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC9A3 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3)
Clonality: monoclonal
Clone: 10A8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: