Anti-TNC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4C8MS]
Catalog # NOVUNB110-68136H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Tenascin C
- Antigen symbol:TNC
- Clonality:Monoclonal
- Clone:4C8MS
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:3371
- Antigen synonyms:HXB|JI|TN|GMEM|150-225|GP|DFNA56|TN-C
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TNC (tenascin C)
Clonality: monoclonal
Clone: 4C8MS
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat