Order Entry
Denmark
ContactUsLinkComponent
Anti-Napsin-A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TMU-Ad02]
Anti-Napsin-A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TMU-Ad02]
Catalog # NOVUNB110-68133F
Supplier:  Novus Biologicals
Anti-Napsin-A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TMU-Ad02]
Catalog # NOVUNB110-68133F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Napsin A
  • Antigen symbol:
    NAPSA
  • Clonality:
    Monoclonal
  • Clone:
    TMU-Ad02
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    9476
  • Antigen synonyms:
    NAPA|Kdap|KAP|NAP1|SNAPA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: Napsin-A
Clonality: monoclonal
Clone: TMU-Ad02
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human