Anti-Napsin-A Mouse Monoclonal Antibody (DyLight® 650) [clone: TMU-Ad02]
Catalog # NOVUNB110-68133C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Napsin A
- Antigen symbol:NAPSA
- Clonality:Monoclonal
- Clone:TMU-Ad02
- Conjugation:DyLight® 650
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:9476
- Antigen synonyms:NAPA|Kdap|KAP|napsin-1|NAP1|TA01/TA02|kidney-derived aspartic protease-like protein|asp 4|aspartyl protease 4|SNAPA|pronapsin A|napsin-A|ASP4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Napsin-A
Clonality: monoclonal
Clone: TMU-Ad02
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human