Anti-STIM1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB110-60547G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Stromal interaction molecule 1
- Antigen symbol:STIM1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:6786
- Antigen synonyms:STRMK|IMD10|TAM|GOK|D11S4896E|TAM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: STIM1 (stromal interaction molecule 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: