Order Entry
Denmark
ContactUsLinkComponent
Anti-APOE Mouse Monoclonal Antibody (DyLight® 550) [clone: WUE-4]
Anti-APOE Mouse Monoclonal Antibody (DyLight® 550) [clone: WUE-4]
Catalog # NOVUNB110-60531R
Supplier:  Novus Biologicals
Anti-APOE Mouse Monoclonal Antibody (DyLight® 550) [clone: WUE-4]
Catalog # NOVUNB110-60531R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein E
  • Antigen symbol:
    APOE
  • Clonality:
    Monoclonal
  • Clone:
    WUE-4
  • Conjugation:
    DyLight® 550
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    348
  • Antigen synonyms:
    apo-E|AD2|LDLCQ5|LPG|MGC1571
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: WUE-4
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse