Order Entry
Denmark
ContactUsLinkComponent
Anti-APOE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: WUE-4]
Anti-APOE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: WUE-4]
Catalog # NOVUNB110-60531F
Supplier:  Novus Biologicals
Anti-APOE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: WUE-4]
Catalog # NOVUNB110-60531F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    apolipoprotein E
  • Antigen symbol:
    APOE
  • Clonality:
    Monoclonal
  • Clone:
    WUE-4
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    348
  • Antigen synonyms:
    APO-E|AD2|LDLCQ5|LPG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: WUE-4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse