Anti-BAP1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 1G8]
NOVUNB110-60521V
- Antibody type:Primary
- Antigen name:BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase)
- Antigen symbol:BAP1
- Clonality:Monoclonal
- Clone:1G8
- Conjugation:DyLight® 405
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:8314
- Antigen synonyms:HUCEP-13|hucep-6|UCHL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 1G8
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human