Anti-CITED4 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HT13-2D6.3]
Catalog # NOVUNB110-41572H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 4
- Antigen symbol:CITED4
- Clonality:Monoclonal
- Clone:HT13-2D6.3
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:163732
- Antigen synonyms:cbp/p300-interacting transactivator 4|MRG-2|transcriptional co-activator 4|MSG1-related protein 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone: HT13-2D6.3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human