Anti-LOX propeptide Rabbit Polyclonal Antibody (DyLight 650)
Catalog # NOVUNB110-41568C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:LOX propeptide
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4015
- Storage buffer:50 mM Sodium Borate
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal LOX propeptide Antibody [DyLight 650]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate, Porcine.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250
Type: Primary
Antigen: LOX propeptide
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human