Anti-GPIHBP1 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB110-41539B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:GPI-anchored HDL-binding protein 1
- Antigen symbol:GPIHBP1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:338328
- Antigen synonyms:GPI-anchored HDL-binding protein 1|GPI-HBP1|1110002J19Rik|GlycosylPhosphatidylInositol-anchored high-de|GPI-anchored High-Density Lipid-binding protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GPIHBP1 (glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse