Anti-ALPP Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 8B6]
Catalog # NOVUNB110-3638F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:alkaline phosphatase, placental
- Antigen symbol:ALPP
- Clonality:Monoclonal
- Clone:8B6
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Gene ID:250
- Antigen synonyms:PALP|ALP|PLAP|PLAP-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ALPP (alkaline phosphatase, placental)
Clonality: monoclonal
Clone: 8B6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human