Order Entry
Denmark
ContactUsLinkComponent
Anti-ALPP Mouse Monoclonal Antibody [clone: 8B6]
Anti-ALPP Mouse Monoclonal Antibody [clone: 8B6]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-ALPP Mouse Monoclonal Antibody [clone: 8B6]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-3638SS
  • NOVUNB110-3638
  • Antigen symbol:
  • ALPP
  • ALP
  • Antigen name:
  • alkaline phosphatase, placental
  • Alkaline Phosphatase
  • Antigen synonyms:
  • PALP|ALP|PLAP|PLAP-1
  • placental alkaline phosphatase 1|AP|Regan isozyme|alkaline phosphatase Regan isozyme|alkaline phosphatase|alkaline phosphomonoesterase|PLAP-1|glycerophosphatase|placental type
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 8B6
  • 8B6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 250
  • 250
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together