Anti-TrkC Rabbit Polyclonal Antibody
Catalog # NOVUNB100-98829
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Neurotrophic Tyrosine Kinase Receptor type 3
- Antigen symbol:TRKC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:4916
- Antigen synonyms:Trk-C|TrkC tyrosine kinase|Neurotrophic tyrosine kinase receptor type 3|NT-3 growth factor receptor|NTRK3|GP145-TrkC
- Storage buffer:Whole antisera
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
The TrkC Antibody from Novus Biologicals is a rabbit polyclonal antibody to TrkC. This antibody reacts with human, mouse, rat. The TrkC Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:300 to 1:2000
Type: Primary
Antigen: TrkC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat