Order Entry
Denmark
ContactUsLinkComponent
 
Anti-CTGF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB100-724H
Supplier:  Novus Biologicals
Anti-CTGF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB100-724H
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Connective Tissue Growth Factor
  • Antigen symbol:
    CTGF
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    1490
  • Antigen synonyms:
    Insulin-like Growth Factor-Binding Protein 8,|IGFBP8|Hcs24|CCN2|Hypertrophic chondrocyte-specific gene product 24|Connective Tissue Growth Factor
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat