Anti-TUBA Mouse Monoclonal Antibody (DyLight® 650) [clone: DM1A]
Catalog # NOVUNB100-690C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Tubulin alpha
- Antigen symbol:TUBA
- Clonality:Monoclonal
- Clone:DM1A
- Conjugation:DyLight® 650
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Gene ID:7846
- Antigen synonyms:alpha Tubulin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: