Anti-6X His tag Mouse Monoclonal Antibody (DyLight® 350) [clone: AD1.1.10]
Catalog # NOVUNB100-64768UV
Supplier: Novus Biologicals
Specifications
- Antibody type:Tag
- Antigen name:His Tag
- Antigen symbol:6xHis
- Clonality:Monoclonal
- Clone:AD1.1.10
- Conjugation:DyLight® 350
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Multiple
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pan