Anti-BRCA1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MU]
Catalog # NOVUNB100-600PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:breast cancer 1, early onset
- Antigen symbol:BRCA1
- Clonality:Monoclonal
- Clone:MU
- Conjugation:PE (Phycoerythrin)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:672
- Antigen synonyms:BROVCA1|PPP1R53|IRIS|FANCS|RNF53|PNCA4|PSCP|BRCAI|BRCC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MU
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human