Anti-BRCA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: KEN]
Catalog # NOVUNB100-599F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:breast cancer 1, early onset
- Antigen symbol:BRCA1
- Clonality:Monoclonal
- Clone:KEN
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:672
- Antigen synonyms:BROVCA1|PPP1R53|IRIS|FANCS|RNF53|PNCA4|PSCP|BRCAI|BRCC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: KEN
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human