Anti-WNVM Rabbit Polyclonal Antibody
NOVUNB100-56743
- Antibody type:Primary
- Antigen name:West Nile Virus Envelope Glyco M
- Antigen symbol:WNVM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:West Nile Virus glycoprotein M
- Storage buffer:PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
The WNVM Antibody from Novus Biologicals is a rabbit polyclonal antibody to WNVM. This antibody reacts with human, mouse, hamster, monkey, virus. The WNVM Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1.0 ?g/ ml
Type: Primary
Antigen: WNVM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Hamster, Monkey, Virus