Anti-TLR4 Mouse Monoclonal Antibody [clone: HTA125]
Catalog # NOVUNB100-56723
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 4
- Antigen symbol:TLR4
- Clonality:Monoclonal
- Clone:HTA125
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Neutralisation:Yes
- Gene ID:7099
- Antigen synonyms:ARMD10, CD284, TLR-4, TOLL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: monoclonal
Clone: HTA125
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: