Order Entry
Denmark
ContactUsLinkComponent
Anti-Anthrax LF Rabbit Polyclonal Antibody
Anti-Anthrax LF Rabbit Polyclonal Antibody
  NOVUNB100-56668
undefined
Anti-Anthrax LF Rabbit Polyclonal Antibody
  NOVUNB100-56668

 

  • Antibody type:
    Primary
  • Antigen name:
    Anthrax Toxin Lethal Factor
  • Antigen symbol:
    ATLF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    Anthrax lethal factor|Anthrax lethal toxin endopeptidase component|LF|Anthrax LF|LEF_BACAN.|bacillus anthracis lethal factor|Lef
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µL
  • Pk:
    0,2 mL

 

 

Type: Primary
Antigen: Anthrax LF
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human