Anti-MBD1 Mouse Monoclonal Antibody [clone: 100B272.1]
Catalog # NOVUNB100-56537
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Methyl-CpG binding domain protein 1
- Antigen symbol:MBD1
- Clonality:Monoclonal
- Clone:100B272.1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:4152
- Antigen synonyms:CXXC3|PCM1|RFT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: MBD1 (methyl-CpG binding domain protein 1)
Clonality: monoclonal
Clone: 100B272.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human