Anti-CVID Rabbit Polyclonal Antibody
NOVUNB100-56516
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 13B
- Antigen symbol:TNFRSF13B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:23495
- Antigen synonyms:CD267|CVID|RYZN|TACI|IGAD2|CVID2|TNFRSF14B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: CVID
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse