Anti-SMAD6 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56440
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:SMAD family member 6
- Antigen symbol:SMAD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4091
- Antigen synonyms:MADH7|HsT17432|MADH6|SMAD6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SMAD6 (SMAD family member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: