Anti-HIST4H4 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56348
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:histone cluster 4, H4
- Antigen symbol:HIST4H4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:121504
- Antigen synonyms:H4/p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human