Anti-FLT1 Rabbit Polyclonal Antibody (Biotin) [clone: 4C11]
Catalog # NOVUNB100-527B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:FMS-related tyrosine kinase 1
- Antigen symbol:FLT1
- Clonality:Polyclonal
- Clone:4C11
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:2321
- Antigen synonyms:FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone: 4C11
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human