Order Entry
Denmark
ContactUsLinkComponent
 
Anti-AGGF1 Rabbit Polyclonal Antibody
 
undefined
Anti-AGGF1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB100-454
  • NOVUNB100-454SS
  • Antigen symbol:
  • VG5Q
  • VG5Q
  • Antigen name:
  • Angiogenic factor with G patch And FHA Domains 1
  • Angiogenic factor with G patch And FHA Domains 1
  • Antigen synonyms:
  • angiogenic factor with G-patch and FHA domains 1|HSU84971|GPATCH7|HUS84971|VG5Q|GPATC7|AGGF1
  • angiogenic factor with G-patch and FHA domains 1|HSU84971|GPATCH7|HUS84971|VG5Q|GPATC7|AGGF1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 55109
  • 55109
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together