Order Entry
Denmark
ContactUsLinkComponent
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: KC21]
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: KC21]
Supplier:  Novus Biologicals
Certificates
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: KC21]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-406SS
  • NOVUNB100-406
  • Antigen symbol:
  • 14-3-3 gamma
  • 14-3-3 gamma
  • Antigen name:
  • Protein kinase C inhibitor protein 1
  • Protein kinase C inhibitor protein 1
  • Antigen synonyms:
  • KCIP-1
  • KCIP-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • KC21
  • KC21
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 7532
  • 7532
  • Isotype:
  • IgG2a
  • IgG2a
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together