Anti-ATM Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10H11.E12]
Catalog # NOVUNB100-306H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Ataxia telangiectasia mutated
- Antigen symbol:ATM
- Clonality:Monoclonal
- Clone:10H11.E12
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:472
- Antigen synonyms:AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: 10H11.E12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat