Anti-KRT1 Mouse Monoclonal Antibody (DyLight® 755) [clone: LHK1]
Catalog # NOVUNB100-2756IR
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:keratin 1, type II
- Antigen symbol:KRT1
- Clonality:Monoclonal
- Clone:LHK1
- Conjugation:DyLight® 755
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Gene ID:3848
- Antigen synonyms:KRT1A|NEPPK|EHK|CK1|K1|EHK1|EPPK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: monoclonal
Clone: LHK1
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat