Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: PyMT]
NOVUNB100-2749F
- Antibody type:Primary
- Antigen name:Polyoma Virus, Medium T Antigen
- Clonality:Monoclonal
- Clone:PyMT
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rat
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Virus
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: Polyoma Virus, Medium T Antigen
Clonality: monoclonal
Clone: PyMT
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Virus