Order Entry
Denmark
ContactUsLinkComponent
Anti-RNF5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LHM 2]
Anti-RNF5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LHM 2]
Catalog # NOVUNB100-2688PE
Supplier:  Novus Biologicals
Anti-RNF5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LHM 2]
Catalog # NOVUNB100-2688PE
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chondroitin sulfate proteoglycan 4
  • Antigen symbol:
    CSPG4
  • Clonality:
    Monoclonal
  • Clone:
    LHM 2
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    1464
  • Antigen synonyms:
    MEL-CSPG|HMW-MAA|MCSP|MCSPG|NG2|MSK16
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: LHM 2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human